Would you like to log in again?
Mayo Trays, Perforated, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10-1687-6 10-1685 10-1689-6 10-1685-12 10-1687 10-1684-6 10-1684-12 10-1686-12 10-1685-6 10-1688 10-1688-12 10-1684 10-1688-6 10-1687-12 10-1686 10-1686-6
10193-132PK 2555.69 USD InStock
10193-132 10193-186 10193-142 10193-130 10193-140 10161-800 10193-190 10163-446 10193-136 10193-124 10193-146 10193-134 10193-144 10193-128 10193-138 10193-126
Mayo Trays, Perforated, Sklar®
Trays/Dishes/Pots Instrument Trays/Dishes
The Mayo Tray holds instruments to be used in a surgical procedure
It is typically organized and monitored by the scrub person or surgical technologist assisting in the procedure. It is used in conjunction with a stand so that the tray and instruments are situated above the waist and easily accessible during the procedure. The tray holds instruments that are the most frequently used during any given procedure. The tray is available perforated or non perforated and in a variety of sizes. Stainless steel material.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty