Would you like to log in again?
Micro Trays, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10-1708 10-1707 10-1716 10-1709 10-1702
76191-446EA 1794.04 USD InStock
76191-446 10193-230 10193-238 10193-248 10193-236
Micro Trays, Sklar®
Trays/Dishes/Pots Instrument Trays/Dishes
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Micro Sterilizing Trays are used to hold smaller instruments securely during a sterilization process. Rows of separating pins and silicone hold down bars for locking instruments in place. The tray also has a hinged lid, locking latch, and outside handles.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty