Would you like to log in again?
Crown Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
24-2346 24-2341 24-2340 24-2345
10193-294EA 102.68 USD InStock
10193-294 10195-418 10192-234 10195-416
Crown Scissors, OR Grade, Sklar
Scissors
Sklar's® Crown Scissors are small finger ring scissors commonly used in dental procedures for trimming crowns
Censitrac Ready products are industry standard and constructed of stainless steel.
They have short jaws for making small cuts when sizing polycarbonate, stainless steel, or aluminum crowns
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty