Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mini Tischler Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-7530
10193-306EA 1533.45 USD InStock
10193-306
Mini Tischler Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Mini Tischler Biopsy Punch Forceps are an OB/GYN instrument used in colposcopy procedures to obtain cervical tissue samples for biopsy.
This product is straight with an 8" shaft and 2.3 x 4.2-mm semi elliptical bite
It features a single, lower tooth used to grasp difficult to reach tissue and a locking mechanism above the horn on the handle, so the jaws can lock the tissue sample into place while the instrument is removed from the cervical canal. This reduces the risk of losing the tissue sample as the instrument is withdrawn. This product has a pistol style handle with one large ring for multiple fingers.
The mini Tischler bite is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred, because there is less bleeding and pain when the smaller tissue sample is removed.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty