Would you like to log in again?
Bruns Bone Curette, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40-7100 40-7204 40-7111 40-7118 40-7116 40-7120 40-7200 40-7112 40-7201 40-7114 40-7113 40-7110 40-7115 40-7000
10196-200EA 187.12 USD InStock
10196-200 10196-214 10196-202 10196-196 10193-350 10196-198 10196-208 10194-236 10192-486 10192-268 10196-206 10193-814 10192-480 10193-786
Bruns Bone Curette, OR Grade, Sklar
Curettes
Bruns Bone Curette is a debulking instrument commonly used in orthopedic procedures and neurosurgery. Also known as a Spratt Bone Curette. It is used to scrape away tissue and bone from the spine, bones, and joints.
  • Reusable and latex-free
  • Industry standard
  • Premium OR-grade
  • Non sterile
It is available in multiple sizes with various curvatures and tip styles to suit the surgical site and specifics of the procedure
This curette has a large, lightweight, hollow handle with longitudinal grooves and a thumb/finger rest for increased control.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty