Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Hartmann Alligator Ear Forceps, OR Grade, Sklar
Supplier: Sklar
67-1021 67-1050 67-1036 67-1020 67-1040 67-1035 67-1039 67-1045 67-1038
10194-160EA 933.18 USD InStock
10194-160 10198-274 10198-120 10193-362 10194-042 10193-300 10193-860 10194-158 10194-136
Hartmann Alligator Ear Forceps, OR Grade, Sklar
Forceps
Hartmann Alligator Forceps are an ENT instrument used in myringotomy procedures to insert or remove tubes or dressings and packing.
  • This product has 3.5" shanks that angle at a screw lock to a 2.5" shaft and 8mm long triangular tips. The tips are horizontally serrated and each have a central groove to securely grip surgical materials, as well as granulation tissue and cerumen.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty