Would you like to log in again?
Merit™ Iris Dressing Forceps, Physician Grade, Sklar
Supplier: Sklar
Merit™
98-123
10193-652EA 23.28 USD InStock
10193-652
Merit™ Iris Dressing Forceps, Physician Grade, Sklar
Forceps
Merit™ Iris Dressing Forceps are thumb style forceps used for placing or removing dressings in many types of procedures
Serrated jaws assist in a having a secure hold on items.
Merit™ instruments are office-grade products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.
Although Dressing Forceps were designed to hold materials such as gauze, they have come to have a variety of additional functions. They may be used for removing foreign objects or dead tissue from wounds, as well as other small surgical materials such as tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty