Would you like to log in again?
Merit™ Metzenbaum Scissors, Physician Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Merit™
97-335 97-340 97-336 97-339
10184-410EA 21.73 USD InStock
10184-410 10184-412 10193-658 10184-414
Merit™ Metzenbaum Scissors, Physician Grade, Sklar
Scissors
Merit™ Metzenbaum dissecting scissors are used for dissecting and cutting fine tissue.
Metzenbaum scissors are identifiable by their long, thin shanks and short jaws with fine, blunt, rounded tips. These scissors are not recommended for cutting sutures, drains, or heavy tissue.
Merit™ instruments are office-grade line of products used for routine and in-office minor procedures. These high quality instruments are crafted from Pakistan steel and come with a ten year guarantee. Merit™ instruments deliver a solution for physicians and practices, that require quality and durable instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty