Would you like to log in again?
Collier Needle Holder, OR Grade, Sklar
Supplier: Sklar
65-6550
10193-718EA 208.95 USD InStock
10193-718
Collier Needle Holder, OR Grade, Sklar
Forceps
Sklar's Collier Needle Holder is a ratcheted, finger ring instrument used for holding needles
Censitrac Ready products are industry standard and constructed of stainless steel.
This device is straight with fenestrated, serrated tips.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty