Would you like to log in again?
Probe with Eye, OR Grade, Sklar
Supplier: Sklar
30-1950 30-2012 30-1955 30-1960
10192-262EA 25.05 USD InStock
10192-262 10193-330 10193-740 10192-416
Probe with Eye, OR Grade, Sklar
Dissection Needles
Probe with Eye is a slender, malleable, multipurpose instrument with a bulbous tip that can be used in a variety of procedures.
It may be used to explore or gauge the depth of non-visible cavities, explore lesions, or manipulate tissue. The eye may be used for carrying ligatures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty