Would you like to log in again?
Foerster Sponge Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
87-2095 87-2395 87-2490 87-2295 87-2195
10015-652EA 302.92 USD InStock
10015-652 10193-660 10199-044 10199-040 10193-764
Foerster Sponge Forceps, OR Grade, Sklar
Forceps
Censitrac Ready products are industry standard and constructed of stainless steel.
  • Foerster Sponge Forceps, also known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster sponge forceps are often used in bariatric surgeries because of their longer length, and the curved version may be used for removing the placenta in OB/GYN procedures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty