Would you like to log in again?
Magill Catheter Forceps, OR Grade, Sklar
Supplier: Sklar
07-1777 07-1780 07-1797
10194-712EA 327.47 USD InStock
10194-712 10193-662 10193-776
Magill Catheter Forceps, OR Grade, Sklar
Forceps
Magill Catheter Forceps are used for grasping endotracheal tube during intubation and removing foreign bodies from the airway or other types of catheters during insertion.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty