Would you like to log in again?
Schamberg Comedone Extractor, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
06-4120 06-4121 06-4122
10193-808EA 39.42 USD InStock
10193-808 10194-896 10194-898
Schamberg Comedone Extractor, OR Grade, Sklar
Curettes
Sklar's Schamberg Comedone Extractor is a dermal instrument used to treat facial blemishes.
It is a double-ended instrument with a small wire loop on one end and a larger, flattened wire loop on the other. Gentle pressure is applied to the appropriate loop end when placed around the blemish, and the instrument assists in forcing the debris causing the blemish out from beneath the skin. The Schamberg Comedone Extractor is available in three varieties, tailored to treat various size blemishes.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty