Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lucae Bayonet Forceps, OR Grade, Sklar
Supplier: Sklar
67-1555 67-1455
10193-824EA 66.13 USD InStock
10193-824 10194-044
Lucae Bayonet Forceps, OR Grade, Sklar
Forceps
Lucae Bayonet Forceps are thumb forceps used for intra-aural procedures.
  • They may be used for procedures in the nasal cavity
  • The offset bayonet handle provides an unobstructed view of confined surgical sites
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty