Would you like to log in again?
Pederson Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-3712 90-3713 90-3851 90-3711 90-3849 90-3710
10193-804EA 79.29 USD InStock
10193-804 10193-948 10193-664 10197-238 10193-852 10193-698
Pederson Vaginal Speculum, OR Grade, Sklar
Specula
Sklar® Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed.
  • The blades are then opened by an external lever and screw
  • Reusable and latex-free
The blades of the Pederson speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty