Would you like to log in again?
Merit™ Mayo-Hegar Needle Holders, OR Grade, Sklar®
Supplier: Sklar
97-847 97-842 97-869 97-853
10193-516EA 26.95 USD InStock
10193-516 10184-924 10193-892 10193-720
Merit™ Mayo-Hegar Needle Holders, OR Grade, Sklar®
Forceps
The Merit™ Mayo-Hegar Needle Holder is an office-grade, ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures
Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty