Would you like to log in again?
Heaney Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-4682
10193-962EA 265.12 USD InStock
10193-962
Heaney Needle Holder, OR Grade, Sklar
Forceps
Sklar's Heaney Needle Holder is a ratcheted, finger ring instrument used for light to medium weight suturing
It is most commonly used in gynecological procedures such as vaginal and abdominal hysterectomy, and dilation and curettage. The slightly curved shanks and tips provide the user a better view when suturing intravaginally, and the cross-serrations provide a secure hold on the needle. This needle holder may also be used as a cardiovascular approximation and closure instrument.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty