Would you like to log in again?
Spear Point Comedone Extractor, OR Grade, Sklar
Supplier: Sklar
06-4126
10193-978EA 42.88 USD InStock
10193-978
Spear Point Comedone Extractor, OR Grade, Sklar
Dissection Needles
Sklar's Spear Point Comedone Extractor is a dermal instrument used to treat facial blemishes.
  • Double ended
This instrument features one sharp, spear-shaped end for use in rupturing pustules and a round, cup-shaped end with a small round hole specifically designed for extraction. Gentle pressure is applied to the appropriate end when the round opening is placed around the blemish, and the instrument assists in forcing the debris causing the blemish out from under the skin.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty