Would you like to log in again?
Sims Uterine Curette Set, OR Grade, Sklar
Supplier: Sklar
90-5428
10194-002PK 1070.56 USD InStock
10194-002
Sims Uterine Curette Set, OR Grade, Sklar
Curettes
Sims uterine curette set is a collection of 6 OB/GYN curettes used for removing tissue from the lining of the uterus.
  • Stainless steel
They may be used to scrape and remove growths or tissue to be biopsied. They may also be used as part of a D and C procedure. The teardrop shaped, fenestrated, loop tip is sharp and available in six sizes in this set. The shaft is malleable so that it can be manipulated to an angle suitable for both user and patient needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty