Would you like to log in again?
Harrington-Mayo Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
55-8611 55-8711
10194-020EA 520.53 USD InStock
10194-020 10197-668
Harrington-Mayo Dissecting Scissors, OR Grade, Sklar
Scissors
Harrington-Mayo Dissecting Scissors feature the bevelled edge of Mayo Dissecting Scissors with longer shanks
Available straight or curved, they are used for dissection in urologic, genito urinary, gynecological and thoracic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty