Would you like to log in again?
Kevorkian Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-7540
10194-070EA 1541.67 USD InStock
10194-070
Kevorkian Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Kevorkian Biopsy Punch is used during colposcopy procedures to excise tissue from the endocervix for biopsy.
This product has a 3 x 8-mm rectangular bite with 4 lower teeth. There is a basket to catch the tissue sample after excision. The non-ratcheted, spring style handle features one large ring that can fit multiple fingers and has a horn on the upper handle for thumb placement.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty