Would you like to log in again?
Sklar Blue™ Potts-Smith Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
91-5413
10194-078EA 377.22 USD InStock
10194-078
Sklar Blue™ Potts-Smith Tissue Forceps, OR Grade, Sklar
Forceps
Blue™ Potts-Smith Tissue Forceps are used for grasping fine or delicate tissue in electrosurgical procedures
Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.
Sklar Blue™ instruments are autoclavable and may be cleaned ultrasonically.
These narrow forceps grasp the tissue, but also have a stop peg to prevent a harsh grasp that may crush the tissue.
Certifications: All Sklar Blue™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Blue™ electrosurgical coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty