Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-8962 90-8960 90-8972
10184-200EA 1233.66 USD InStock
10184-200 10184-194 10194-154
Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Micro-Cut Universal Style Biopsy Punch Forceps are an OB/GYN instrument used in colposcopy procedures to obtain cervical tissue samples for biopsy
The oval bite is 3mm x 5mm, which is slightly smaller than a standard Tischler bite and slightly larger than a Mini Tischler bite. Small punches are often preferred because there is typically less bleeding and pain; however, there are circumstances in which a larger sample is necessary. This punch provides a size in between standard options. This product is a complete instrument which consists of a handle and a detachable cannula/tip so other universal tip styles may be used with one handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty