Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Merit™ Vienna Nasal Speculum, Physician Grade, Sklar
Supplier: Sklar
98-268 98-267 98-258
10194-208EA 82.4 USD InStock
10194-208 10185-294 10185-292
Merit™ Vienna Nasal Speculum, Physician Grade, Sklar
Specula
Intranasal instrument used to retract and expose nasal tissue for examination.
  • This also known as Wien model
Most commonly utilized for routine and in-office minor procedures. These cost effective, high-quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty