Would you like to log in again?
Neivert Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-1760
10194-268EA 236.66 USD InStock
10194-268
Neivert Needle Holder, OR Grade, Sklar
Forceps
Neivert needle holder ensures a strong, anti-slip grip via its serrated jaws
Neivert needle holders are industry standard and constructed of stainless steel.
The angled ring handle eases comfort for the surgeon.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty