Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potts-Smith Scissors, OR Grade, Sklar

Supplier: Sklar

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

52-2860 98-1075 52-2825 52-2845 98-1076
10199-416EA 422.08 USD InStock
10199-416 10194-308 10184-952 10184-954 10194-314
Potts-Smith Scissors, OR Grade, Sklar
Scissors
Sklar's® Potts-Smith Scissors are a finger ring instrument commonly used for trimming and opening vessels in cardiovascular and thoracic procedures.

Blades are available in a 20 mm intermediate weight or 13 mm delicate weight style

Censitrac Ready products are industry standard and constructed of stainless steel.

Delicate Potts-Smith scissors are heavier than Diethrich scissors, and can therefore also be used for cutting through plaque.

Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.

Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty