Would you like to log in again?
Douglas Cilia Forceps, OR Grade, Sklar
Supplier: Sklar
66-1830
10194-386EA 60.35 USD InStock
10194-386
Douglas Cilia Forceps, OR Grade, Sklar
Forceps
Douglas Cilia Forceps are ophthalmologic thumb forceps used for removing eyelashes, often as part of treatment for blepharitis marginalis.
This product has 2.5-mm squared tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty