Would you like to log in again?
Bergh Cilia Forceps, OR Grade, Sklar
Supplier: Sklar
66-1635
10194-388EA 78.83 USD InStock
10194-388
Bergh Cilia Forceps, OR Grade, Sklar
Forceps
Bergh Cilia Forceps are heavy spring thumb forceps used during ophthalmologic procedures involving the eyelid, commonly, blepharitis marginalis.
These forceps are serrated and used for grasping and removing cilia, or eyelashes. The flat, grooved handle provides a secure grip for the user.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty