Would you like to log in again?
Francis Chalazion Forceps, OR Grade, Sklar
Supplier: Sklar
66-1255
10194-394EA 419.09 USD InStock
10194-394
Francis Chalazion Forceps, OR Grade, Sklar
Forceps
Francis Chalazion Forceps are ophthalmologic thumb forceps, used to evert the eyelid, and expose chalazion for drainage and removal.
This instrument has two oval tips: One is solid and the other is fenestrated. The chalazion on the eyelid is isolated between the two tips and the forceps are tightened by an attached screw lock, which pushes the chalazion upwards so the procedure can be completed. The size and consistency of the chalazion are primary factors when choosing which chalazion forceps to use. Larger chalazion, as well as those with hard matter will require larger incisions.
Therefore, forceps with larger tips may be useful. Smaller chalazion, or those determined to contain only fluid, may be excised with smaller incisions and the use of forceps with a smaller diameter. The top arm of this product angles upwards immediately after the screw lock, and then angle back down towards the fenestrated ring. The diameter of the tips of this product is approximately 12 x 14-mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty