Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Laufe Polyp Forceps, OR Grade, Sklar
Supplier: Sklar
90-7977
10194-428EA 335.53 USD InStock
10194-428
Laufe Polyp Forceps, OR Grade, Sklar
Forceps
Sklar's® Laufe Polyp Forceps are ratcheted, finger ring forceps commonly used to remove polyps from the uterus
Fenestrated, serrated, oval tips provide an ideal grasping surface for successful removal of these growths. The jaws are curved slightly to provide the user with a view unobstructed by the instrument.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty