Would you like to log in again?
McIndoe Forceps, OR Grade, Sklar
Supplier: Sklar
19-1230
10194-460EA 73.07 USD InStock
10194-460
McIndoe Forceps, OR Grade, Sklar
Forceps
McIndoe Forceps are used to hold tissue, remove debris from wounds, place and remove dressings and packing.
They are non-toothed, thumb forceps with a stop peg to ensure that when used for holding tissue the grip is not crushing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty