Would you like to log in again?
Sims Vaginal Retractor, Single-Ended, OR Grade, Sklar
Supplier: Sklar
90-3225 90-3327 90-3335 90-3230 90-3330 90-3235
10194-472EA 281.29 USD InStock
10194-472 10199-224 10194-474 10199-222 10199-220 10194-476
Sims Vaginal Retractor, Single-Ended, OR Grade, Sklar
Retractors
Sklar's Sims Vaginal Retractor is a handheld retractor used primarily for vaginal retraction and exposure
This retractor is a single-ended instrument with the blades always curved at right angles. The handle is not distinctly separate from the working end of the instrument and is essentially a continuous piece of stainless steel. There is a slight curve at the base of the handle for finger placement. The blade is inserted into the vagina to retract the anterior wall so that the posterior wall can be viewed. The blade is trough shaped, which serves to catch and drain blood or fluid that may exit the surgical site.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty