Would you like to log in again?
Sklarlite XD™ Strabismus Scissors, OR Grade, Sklar®
Supplier: Sklar
23-1162 23-1163
10194-526EA 293.5 USD InStock
10194-526 10194-524
Sklarlite XD™ Strabismus Scissors, OR Grade, Sklar®
Scissors
Sklarlite XD™ Strabismus Scissors are an ophthalmic dissecting instrument primarily used during procedures for the treatment of Strabismus, otherwise known as "cross-eyes." Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue
These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty