Would you like to log in again?
Sklarlite XD™ Littauer Stitch Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
23-1195
10194-536EA 333.2 USD InStock
10194-536
Sklarlite XD™ Littauer Stitch Scissors, OR Grade, Sklar®
Scissors
Sklarlite XD™ Littauer Stitch Scissors are primarily used for suture removal
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Littauer Stitch Scissors are most commonly available in the length of 5-1/2 inches, therefore, they typically have larger, less delicate tips than Spencer Stitch Scissors, and are used for removing larger sutures. They should not be used for cutting tissue. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with smooth, blunt/blunt tips and a length of 5-1/2 inches.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty