Would you like to log in again?
Sklarlite XD™ Hartmann Mosquito Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite XD™
23-2500
10194-550EA 210.48 USD InStock
10194-550
Sklarlite XD™ Hartmann Mosquito Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Hartmann Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures
Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Sklarlite XD™ instruments are lighter and more delicate than standard instruments, which allow the surgeon the highest degree of precision with the least amount of fatigue.
Used in plastic and vascular surgeries, Hartmann Mosquito Forceps can also be used in pediatric and ophthalmologic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty