Would you like to log in again?
Castroviejo Corneal Scissors, OR Grade, Sklar
Supplier: Sklar
64-2242 64-2342 64-2442 64-2535
10194-558EA 305.36 USD InStock
10194-558 10194-552 10194-554 10194-556
Castroviejo Corneal Scissors, OR Grade, Sklar
Scissors
Castroviejo Miniature Corneal Scissors are double spring ophthalmologic scissors most commonly used during corneal transplants or corneal trephination (removing a round piece of the cornea).
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty