Would you like to log in again?
Beer Cilia Forceps, OR Grade, Sklar
Supplier: Sklar
66-1770
10194-586EA 148 USD InStock
10194-586
Beer Cilia Forceps, OR Grade, Sklar
Forceps
Beer Cilia Forceps are used for grasping and holding cilia, also known as eyelashes.
The tips of Beer cilia forceps resemble the heel of a shoe. The tip itself is rounded, but the inside working surface, although smooth, is raised very slightly from the rest of the forceps body and creating the look of a shoe's heel.
Beer cilia forceps are similar to Ziegler cilia forceps; however, the tip of the Beer instrument is 4-mm wide, whereas the Ziegler is 2-mm wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty