Would you like to log in again?
Swiss Cilia Suture Forceps, OR Grade, Sklar
Supplier: Sklar
66-1740
10194-588EA 89.55 USD InStock
10194-588
Swiss Cilia Suture Forceps, OR Grade, Sklar
Forceps
Swiss Cilia Suture Forceps are thumb forceps used for removing cilia, foreign bodies, or sutures around the eye and eyelid.
They have slanted, diagonal tips, raised from the rest of the body of the forceps. The tips have fine edges, useful for grasping very small items such as eyelashes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty