Would you like to log in again?
Desmarres Chalazion Forceps, OR Grade, Sklar
Supplier: Sklar
66-1038 66-1036 66-1035
10194-590EA 408 USD InStock
10194-590 10194-592 10194-594
Desmarres Chalazion Forceps, OR Grade, Sklar
Forceps
Desmarres Chalazion Forceps are ophthalmologic thumb forceps used to evert the eyelid and expose chalazion for drainage and removal.
The instrument has two sideways oval tips. One is solid and the other is fenestrated. The chalazion on the eyelid is isolated between the two tips and the forceps are tightened by an attached screw, which pushes the chalazion upwards so a procedure can be completed. The size and consistency of the chalazion are primary factors when choosing which chalazion forceps to use. Larger chalazion, as well as those with hard matter, will require larger incisions. Therefore, forceps with larger tips may be more useful. Similarly, smaller chalazion, or those determined to contain only fluid, may be excised with smaller incisions and the use of forceps with a smaller diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty