Would you like to log in again?
Micro Surgery Needle Holder, OR Grade, Sklar
Supplier: Sklar
98-7820 98-3188 98-3192 98-3185 98-3183 98-3186 98-3182 98-3184 98-3190 98-3180
10194-624EA 1057.55 USD InStock
10194-624 10192-232 75995-892 75818-100 10194-626 10185-312 10194-628 10185-310 10194-620 10194-622
Micro Surgery Needle Holder, OR Grade, Sklar
Forceps
Sklar's® micro needle holder is most commonly used to hold fine suture needles in very small sites or microsurgical procedures.
  • Ergonomical handles are designed with texturized grips for comfort
  • Available in various lengths
Micro needle holder is available in various curvature, straight or curved, with or without a catch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty