Would you like to log in again?
Desmarres Retractor, Smooth, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
65-1843 65-1841 65-1842 65-1844
10194-658EA 141.74 USD InStock
10194-658 10194-660 10194-654 10194-656
Desmarres Retractor, Smooth, OR Grade, Sklar®
Retractors
Desmarres Lid Retractor is a handheld ophthalmologic retractor used for retaining primarily the upper eyelid and eyelashes
It may be used to pull down the lower lid as well. The curved, saddle style blade contours to the round shape of the eye as well as the curvature of the eyelid created when it is retracted. It is available in multiple sizes to fit the patient's eye.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty