Would you like to log in again?
Sklarlite™ Sklarhone™ Metzenbaum Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
15-9109 15-9118 15-9108 15-9107 15-9117
10194-724EA 294.9 USD InStock
10194-724 10194-726 10194-718 10194-720 10194-722
Sklarlite™ Sklarhone™ Metzenbaum Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklarlite™ Sklarhone™ Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only.
These scissors are not recommended for cutting sutures, drains, or heavy tissue. The Sklarlite™ Sklarhone™ combination provides the surgeon with a lightweight, precision instrument that allows the user to make the sharp incisions of a knife with the cutting ability of scissors, all while having greater control and experiencing less fatigue. This instrument is identified by dual black finger rings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty