Would you like to log in again?
Spencer Wells Forceps, OR Grade, Sklar
Supplier: Sklar
17-2560 17-2550 17-2570 17-2580 17-2660 17-2670 17-2678
10194-768EA 77.41 USD InStock
10194-768 10194-760 10194-770 10194-762 10194-772 10194-764 10194-766
Spencer Wells Forceps, OR Grade, Sklar
Forceps
Spencer Wells Forceps are hemostatic forceps used to control blood flow in vessels.
They have short, serrated jaws, which can seal small blood vessels
Censitrac Ready products are industry standard and constructed of stainless steel.
On larger vessels they may be used to temporarily control the blood flow, and left to be sutured later. The handles of the Spencer Wells forceps were designed to be closer together to eliminate tangling of arteries within them. Ratcheted versions only have 1 or 2 locks. They are available with either curved or straight jaws, in a number of lengths.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty