Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Rochester-Ochsner Forceps, OR Grade, Sklar®
Supplier: Sklar
17-2890 17-2690 17-2885 18-1852 17-2685 18-1752 17-2962
10194-778EA 207.93 USD InStock
10194-778 10194-828 10194-782 76192-224 10194-830 10194-774 76192-222
Rochester-Ochsner Forceps, OR Grade, Sklar®
Forceps
Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held
Censitrac Ready OR Grade Rochester-Ochsner Forceps are industry standard.
Rochester-Oschner Forceps are available either curved or straight, in multiple lengths. These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps. Few have horizontal serrations with the entire length of the jaw as well as 1 x 2 teeth at the tip.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty