Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Rochester-Oschner (Kocher) Forceps, OR Grade, Sklar
Supplier: Sklar
17-2855 22-3855C 22-3755 17-2862C 22-3855 17-2762 17-2762C 17-2855C 22-3755C 17-2862
89025-366EA 204.59 USD InStock
89025-366 82030-006 82030-004 75848-088 75846-950 75846-952 75847-192 89025-364 10194-776 75847-190
Rochester-Oschner (Kocher) Forceps, OR Grade, Sklar
Forceps
Rochester-Oschner (Kocher) Forceps are a heavy instrument designed to aggressively grasp medium to heavy tissue or occlude heavy, dense vessels.
  • Autoclavable or latex-free
  • Available either curved or straight, in multiple lengths
  • Forceps have horizontal serrations the entire length of the jaw
  • Serrations plus teeth ensure a firm grip on the tissue or vessel
These forceps are sometimes referred to as simply Oschner Forceps, however, Rochester-Oschner Forceps are actually heavier than standard Oschner Forceps.
Censitrac Ready OR Grade Rochester-Ochsner Forceps are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty