Would you like to log in again?
Lahey Forceps, OR Grade, Sklar
Supplier: Sklar
17-3255 17-3355
10194-792EA 137.86 USD InStock
10194-792 10194-788
Lahey Forceps, OR Grade, Sklar
Forceps
Sklar's® Lahey Forceps are a hemostats commonly used for clamping blood vessels or tissue before cauterization or ligation. They have ratcheted, finger ring handles. Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique.
Lahey’s with shorter jaws may also be used for soft tissue dissection typically no deeper than 6-8 inches, or in laparotomy, thyroid and vaginohysterectomy procedures.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty