Would you like to log in again?
Bainbridge Forceps, OR Grade, Sklar
Supplier: Sklar
17-3762 17-3660
10194-802EA 192.68 USD InStock
10194-802 76192-234
Bainbridge Forceps, OR Grade, Sklar
Forceps
Bainbridge Forceps are ratcheted, finger ring forceps used in intestinal procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
They are longitudinally serrated the entire length of the tapered jaws with cross-serrated tips. Bainbridge Forceps are a light to medium weight, non-crushing instrument, that can hold back the lumen of the intestines, arteries, or organs such as the bowel without causing damage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty