Would you like to log in again?
Barrett Tenaculum Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
18-2257 91-1070
10193-994EA 239.31 USD InStock
10193-994 10194-840
Barrett Tenaculum Forceps, OR Grade, Sklar
Forceps
Barrett Tenaculum Forceps are ratcheted, finger ring forceps used for grasping cervical tissue.
With straight shanks and sharp tips that curve inward, Barrett tenaculum forceps resemble Braun tenaculum forceps; however, Barrett forceps are shorter
Censitrac Ready products are industry standard and constructed of stainless steel.
The sharp, pointed tips ensure a firm grip on the targeted tissue. These forceps may be used for foreign body removal.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty