Would you like to log in again?
Michel Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
19-1950
10194-842EA 145.6 USD InStock
10194-842
Michel Tissue Forceps, OR Grade, Sklar
Forceps
Michel Tissue Forceps are thumb forceps used for applying Michel's clips or skin sutures.
Michel's clips are metal skin sutures used for approximating the edges of a wound to stop bleeding or for closure.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty