Would you like to log in again?
Lane Tissue Forceps, OR Grade, Sklar®
Supplier: Sklar
19-1957 19-1975 19-1955
10194-844EA 226.18 USD InStock
10194-844 10194-846 76192-314
Lane Tissue Forceps, OR Grade, Sklar®
Forceps
Sklar® Lane Tissue Forceps are non-ratcheted forceps used for holding tough tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Lane Tissue Forceps are non-ratcheted thumb forceps used for holding tough tissue such as fascia and cartilage. The handle is smooth and features a slight thumb/finger indentation for greater control.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty